missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Methyldiphenylamine, 99%
Click to view available options
Quantity:
10 g
50 g
Specifications
Melting Point | 25°C to 27°C |
Density | 1.066 |
Boiling Point | 317°C |
Flash Point | 147°C (297°F) |
Refractive Index | 1.635 |
Quantity | 10 g |
Formula Weight | 183.25 |
Percent Purity | 99% |
Chemical Name or Material | 3-Methyldiphenylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'