missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylcinnolin-5-amine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Synonym | 3-methyl-cinnolin-5-ylamine, 5-cinnolinamine, 3-methyl, 3-methyl-5-cinnolinamine, 3-methylcinnoline-5-ylamine, maybridge1_006688, 5-cinnolinamine,3-methyl, cambridge id 5256404, 3-methylcinnolin-5-yl amine, 5-amino-3-methyl-1,2-benzodiazine |
Formula Weight | 159.19 |
Percent Purity | 95% |
Chemical Name or Material | 3-Methylcinnolin-5-amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'