missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylbut-2-enyl formate, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Linear Formula | (CH3)2C=CHCH2OCHO |
Quantity | 1 g |
IUPAC Name | 3-methylbut-2-en-1-yl formate |
Formula Weight | 114.14 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 3-Methylbut-2-enyl formate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'