missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (3-Methylbenzyl)triphenylphosphonium chloride, 98%

Brand:  Alfa Aesar™ A11611.09

Code : NEW

Additional Details : CAS Number : 63368-37-6 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11414703

  • £25.90 / 10g


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Specifications

Chemical Identifiers

(3-Methylbenzyl)triphenylphosphonium chloride
C26H24ClP
MFCD00031579
63368-37-6
402.91
4071111

Specifications

(3-Methylbenzyl)triphenylphosphonium chloride
98%
402.91
4071111
10g
63368-37-6
C26H24ClP
MFCD00031579
>300°C
Videos
SDS