missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Methylbenzyl isothiocyanate, 99%

Brand:  Alfa Aesar™ H54893.06

Code : NEW

Additional Details : CAS Number : 3696-66-0 Weight : 0.00500kg Transport : UN number : 2922 Chem class : 1b Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15461048

  • £233.20 / 5g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Methylbenzyl isothiocyanate
163.24
UN2922
3696-66-0
MFCD00060410

Specifications

3-Methylbenzyl isothiocyanate
1.09g/mL
99%
MFCD00060410
Pale brown
5g
3696-66-0
Liquid
163.24
UN2922
127°C (2mmHg)
Videos
SDS