missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(Methylamino)pyridine, 95%

Brand:  Alfa Aesar™ H63602.06

Code : NEW

Additional Details : CAS Number : 18364-47-1 Weight : 0.00500kg Transport : UN number : 2810 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15470379

  • £282.30 / 5g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

3-(Methylamino)pyridine
108.14
UN2810
18364-47-1
MFCD01418130
3-Methylaminopyridine

Specifications

3-(Methylamino)pyridine
5g
Liquid
108.14
UN2810
Yellow
18364-47-1
95%
MFCD01418130
3-Methylaminopyridine
Videos
SDS