missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3-Methyladenine, 90+%

Catalog Number p-3200943
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass Bottle
5142-23-4
C6H7N5
149.16
MFCD00010531
FSASIHFSFGAIJM-UHFFFAOYSA-N
3-methyladenine, 3-methyl-3h-purin-6-amine, 6-amino-3-methylpurine, 3-ma, 3h-purin-6-amine, 3-methyl, 3-methyl-3h-adenine, n3-methyladenine, adenine, 3-methyl, 3-methyl-3h-purin-6-ylamine, 3-methyladenine 3-ma
1673
CHEBI:38635
CN1C=NC(N)=C2N=CN=C12
5142-23-4
C6H7N5
149.16
MFCD00010531
FSASIHFSFGAIJM-UHFFFAOYSA-N
3-methyladenine, 3-methyl-3h-purin-6-amine, 6-amino-3-methylpurine, 3-ma, 3h-purin-6-amine, 3-methyl, 3-methyl-3h-adenine, n3-methyladenine, adenine, 3-methyl, 3-methyl-3h-purin-6-ylamine, 3-methyladenine 3-ma
1673
CHEBI:38635
CN1C=NC(N)=C2N=CN=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 90.0
CAS Max % 100.0
Melting Point 300°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 90% min. (HPLC)
Packaging Glass Bottle
Quantity 100 mg
Beilstein 26,V,17,151
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol,dmso,chloroform
IUPAC Name 3-methyl-3H-purin-6-amine
Formula Weight 149.15
Percent Purity 90+%
Physical Form Powder
Chemical Name or Material 3-Methyladenine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyladenine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'