missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethyl)pyrazole, 99%, Thermo Scientific™

Catalog Number p-3833378
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10010-93-2
C5H5F3N2
150.1
MFCD00829309
DLCHCAYDSKIFIN-UHFFFAOYSA-N
5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole
139077
5-methyl-3-(trifluoromethyl)-1H-pyrazole
CC1=CC(=NN1)C(F)(F)F
This item is not returnable. View return policy
10010-93-2
C5H5F3N2
150.1
MFCD00829309
DLCHCAYDSKIFIN-UHFFFAOYSA-N
5-methyl-3-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl-1h-pyrazole, 3-methyl-5-trifluoromethyl pyrazole, 1h-pyrazole, 3-methyl-5-trifluoromethyl, 5-methyl-3-trifluoromethyl pyrazole, 3-trifluoromethyl-5-methyl pyrazole, pyrazole, 3-methyl-5-trifluoromethyl, 3-methyl-5-trifluoromethylpyrazole, 3-trifluoromethyl-5-methylpyrazole, 3-methyl-5-trifluoromethyl-2h-pyrazole
139077
5-methyl-3-(trifluoromethyl)-1H-pyrazole
CC1=CC(=NN1)C(F)(F)F
CAS Min % 98.5
CAS Max % 100.0
Melting Point 88.0°C to 90.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 150.1
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Methyl-5-(trifluoromethyl)pyrazole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-5-(trifluoromethyl)pyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'