Learn More
Alfa Aesar™ 3-Methyl-5-(trifluoromethoxy)benzonitrile, 97%
Brand: Alfa Aesar™ H31921.03
Code : NEW
Additional Details : Weight : 0.00100kg Transport : UN number : 3276 Chem class : 6.1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
916420-59-2 | |
201.148 | |
RUVNGFLRCPBNJK-UHFFFAOYSA-N | |
21412932 | |
CC1=CC(=CC(=C1)C#N)OC(F)(F)F |
C9H6F3NO | |
MFCD09025378 | |
3-methyl-5-trifluoromethoxy benzonitrile, 3-cyano-5-trifluoromethoxy toluene, 5-methyl-3-trifluoromethoxy benzenecarbonitrile | |
3-methyl-5-(trifluoromethoxy)benzonitrile |
Specifications
3-Methyl-5-(trifluoromethoxy)benzonitrile | |
1g | |
C9H6F3NO | |
UN3276 | |
RUVNGFLRCPBNJK-UHFFFAOYSA-N | |
3-methyl-5-(trifluoromethoxy)benzonitrile | |
21412932 | |
97% |
1.4545 | |
916420-59-2 | |
MFCD09025378 | |
3-methyl-5-trifluoromethoxy benzonitrile, 3-cyano-5-trifluoromethoxy toluene, 5-methyl-3-trifluoromethoxy benzenecarbonitrile | |
CC1=CC(=CC(=C1)C#N)OC(F)(F)F | |
201.148 | |
201.15 |