missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-isoxazoleacetic acid, 98%, Thermo Scientific™

Catalog Number p-3770974
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19668-85-0
C6H7NO3
141.13
MFCD01863539
POEFJFLAFQWOTL-UHFFFAOYSA-N
2-3-methylisoxazol-5-yl acetic acid, 3-methyl-5-isoxazoleacetic acid, 5-isoxazoleacetic acid, 3-methyl, 2-3-methyl-1,2-oxazol-5-yl acetic acid, 3-methyl-1,2-oxazol-5-yl acetic acid, 5-isoxazoleacetic acid,3-methyl, zlchem 1304, 3-methyl-5-isoxazole acetic acid, 3-methyl-isoxazol-5-yl-acetic acid, 5-carboxymethyl-3-methylisoxazole
4195532
2-(3-methyl-1,2-oxazol-5-yl)acetic acid
CC1=NOC(=C1)CC(=O)O
This item is not returnable. View return policy
19668-85-0
C6H7NO3
141.13
MFCD01863539
POEFJFLAFQWOTL-UHFFFAOYSA-N
2-3-methylisoxazol-5-yl acetic acid, 3-methyl-5-isoxazoleacetic acid, 5-isoxazoleacetic acid, 3-methyl, 2-3-methyl-1,2-oxazol-5-yl acetic acid, 3-methyl-1,2-oxazol-5-yl acetic acid, 5-isoxazoleacetic acid,3-methyl, zlchem 1304, 3-methyl-5-isoxazole acetic acid, 3-methyl-isoxazol-5-yl-acetic acid, 5-carboxymethyl-3-methylisoxazole
4195532
2-(3-methyl-1,2-oxazol-5-yl)acetic acid
CC1=NOC(=C1)CC(=O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 101.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 141.13
Percent Purity 98%
Physical Form Needle-like Crystals or Fine Crystalline Powder
Chemical Name or Material 3-Methyl-5-isoxazoleacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-5-isoxazoleacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'