missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 208.17 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'