missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-oxetanemethanol, 98%, ACROS Organics™

£24.90 - £76.30

Chemical Identifiers

CAS 3143-02-0
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00010273
InChI Key NLQMSBJFLQPLIJ-UHFFFAOYSA-N
Synonym 3-methyl-3-oxetanemethanol, 3-methyloxetan-3-yl methanol, 3-hydroxymethyl-3-methyloxetane, 3-oxetanemethanol, 3-methyl, 3-methyl-3-oxetanyl methanol, 3-methyloxetane-3-methanol, 3-methyl-3-oxethanemethanol, 3-methyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-methyl-oxetane, 3-methyl-oxetan-3-yl-methanol
PubChem CID 137837
IUPAC Name (3-methyloxetan-3-yl)methanol
SMILES CC1(COC1)CO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10451104
SDS View Documents
Acros Organics
240840100
10g Glass bottle
£24.90
10g
View alternative products
Save Up to 
»
Estimated Shipment: 04-10-2021
Please to purchase this item. Need a web account? Register with us today!
10031754
SDS View Documents
Acros Organics
240840500
50g Glass bottle
£76.30
50g
View alternative products
Save Up to 
»
Estimated Shipment: 07-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

3143-02-0
102.13
NLQMSBJFLQPLIJ-UHFFFAOYSA-N
137837
CC1(COC1)CO
C5H10O2
MFCD00010273
3-methyl-3-oxetanemethanol, 3-methyloxetan-3-yl methanol, 3-hydroxymethyl-3-methyloxetane, 3-oxetanemethanol, 3-methyl, 3-methyl-3-oxetanyl methanol, 3-methyloxetane-3-methanol, 3-methyl-3-oxethanemethanol, 3-methyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-methyl-oxetane, 3-methyl-oxetan-3-yl-methanol
(3-methyloxetan-3-yl)methanol

Specifications

3-Methyl-3-oxetanemethanol
97°C
Glass bottle
122.0°C (70.0 mmHg)
10g
97.5
97.5% min. (GC)
MFCD00010273
NLQMSBJFLQPLIJ-UHFFFAOYSA-N
(3-methyloxetan-3-yl)methanol
137837
Liquid
1.0340g/mL
Authentic
1.034
Colorless to Yellow
3143-02-0
100.0
C5H10O2
3-methyl-3-oxetanemethanol, 3-methyloxetan-3-yl methanol, 3-hydroxymethyl-3-methyloxetane, 3-oxetanemethanol, 3-methyl, 3-methyl-3-oxetanyl methanol, 3-methyloxetane-3-methanol, 3-methyl-3-oxethanemethanol, 3-methyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-methyl-oxetane, 3-methyl-oxetan-3-yl-methanol
CC1(COC1)CO
102.13
102.13
98%
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Methyl-3-oxetanemethanol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

SDS
Product Certifications