missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-thiophenecarboxaldehyde, 90%, Tech., ACROS Organics™

Brand:  Acros Organics 172781000

Code : 29

Additional Details : CAS Number : 5834-16-2 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10134000

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3-Methyl-2-thiophenecarboxaldehyde
5834-16-2
100.0
≥88%
C6H6OS
MFCD00005430
tech. 90%
88.0
Technical Grade
90%
126.17

Specifications

3-Methyl-2-thiophenecarboxaldehyde
100g
5834-16-2
100.0
Authentic
Technical Grade
≥88%
C6H6OS
MFCD00005430
Clear yellow to brown
tech. 90%
88.0
1.1700g/mL
Glass bottle
Liquid
90%
126.17
17, III, 4521
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Methyl-2-thiophenecarboxaldehyde

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS