missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pentanone, 99%, Thermo Scientific Chemicals

Catalog Number 11914821
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
565-61-7
C6H12O
100.16
UIHCLUNTQKBZGK-UHFFFAOYSA-N
3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
11262
3-methylpentan-2-one
CCC(C)C(=O)C
This item is not returnable. View return policy
565-61-7
C6H12O
100.16
UIHCLUNTQKBZGK-UHFFFAOYSA-N
3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
11262
3-methylpentan-2-one
CCC(C)C(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.8100g/mL
Boiling Point 118.0°C (758.0 mmHg)
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(CH3)COCH3
Refractive Index 1.4002
Quantity 1 g
Beilstein 01,693
Specific Gravity 0.81
Formula Weight 100.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-pentanone
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-2-pentanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P223-Do not allow contact with water.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'