missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-oxobutanoic acid, sodium salt, 98+%, Thermo Scientific Chemicals

Catalog Number p-166539
36.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3715-29-5
C5H7NaO3
138.1
MFCD00002581
WIQBZDCJCRFGKA-UHFFFAOYSA-M
sodium 3-methyl-2-oxobutanoate, ketovaline sodium salt, unii-5om3h751iu, butanoic acid, 3-methyl-2-oxo-, sodium salt, 3-methyl-2-oxobutyric acid sodium salt, 2-ketoisovaleric acid sodium salt, sodium 3-methyl-2-oxobutyrate, 3-methyl-2-oxobutanoic acid sodium salt, sodium dimethylpyruvate, ketovaline sodium
2724059
sodium;3-methyl-2-oxobutanoate
CC(C)C(=O)C(=O)[O-].[Na+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3715-29-5
C5H7NaO3
138.1
MFCD00002581
WIQBZDCJCRFGKA-UHFFFAOYSA-M
sodium 3-methyl-2-oxobutanoate, ketovaline sodium salt, unii-5om3h751iu, butanoic acid, 3-methyl-2-oxo-, sodium salt, 3-methyl-2-oxobutyric acid sodium salt, 2-ketoisovaleric acid sodium salt, sodium 3-methyl-2-oxobutyrate, 3-methyl-2-oxobutanoic acid sodium salt, sodium dimethylpyruvate, ketovaline sodium
2724059
sodium;3-methyl-2-oxobutanoate
CC(C)C(=O)C(=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 227°C to 231°C
Color Beige to Yellow or White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CH3)2CHCOCO2Na
Quantity 1 g
Beilstein 03, 682
Formula Weight 138.1
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Methyl-2-oxobutanoic acid, sodium salt, 98+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'