missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals

Catalog Number p-4259012
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
107-86-8
C5H8O
84.12
SEPQTYODOKLVSB-UHFFFAOYSA-N
3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
61020
CHEBI:15825
3-methylbut-2-enal
CC(=CC=O)C
This item is not returnable. View return policy
107-86-8
C5H8O
84.12
SEPQTYODOKLVSB-UHFFFAOYSA-N
3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
61020
CHEBI:15825
3-methylbut-2-enal
CC(=CC=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 132.0°C to 133.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=CHCHO
Refractive Index 1.4610 to 1.4630
Quantity 5 mL
Beilstein 01,IV,3464
Merck Index 15,8587
Specific Gravity 0.88
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether
Formula Weight 84.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-butenal
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'