missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Methyl-1-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64548.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15481819

  • £119.78 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Methyl-1-phenyl-2-thiohydantoin
206.26
MFCD03721167
3-Methyl-1-phenyl-2-thioxo-4-imidazolidinone

Specifications

3-Methyl-1-phenyl-2-thiohydantoin
Solid
206.26
3-Methyl-1-phenyl-2-thioxo-4-imidazolidinone
1g
95%
MFCD03721167
Videos
SDS