missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol, ACS, 98.5+%, Thermo Scientific Chemicals

Catalog Number p-7050403
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
123-51-3
C5H12O
88.15
MFCD00002934
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
31260
CHEBI:15837
3-methylbutan-1-ol
CC(C)CCO
This item is not returnable. View return policy
123-51-3
C5H12O
88.15
MFCD00002934
PHTQWCKDNZKARW-UHFFFAOYSA-N
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
31260
CHEBI:15837
3-methylbutan-1-ol
CC(C)CCO

3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 117°C
Density 0.809
Boiling Point 130°C to 132°C
Flash Point 45°C (113°F)
Assay Percent Range 98.5+%
Refractive Index 1.406
Quantity 500 mL
UN Number UN1105
Beilstein 1718835
Merck Index 14,5195
Solubility Information Miscible with water,alcohol,ether,benzene,chloroform,petroleum ether,glacial acetic acid,acetone and oils.
Formula Weight 88.15
Percent Purity ≥98.5%
Grade ACS
Physical Form Liquid
Chemical Name or Material 3-Methyl-1-butanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'