missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-butanol, 99%, anhydrous, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 433870010

Code : 29

Additional Details : CAS Number : 123-51-3 Weight : 0.86000kg Transport : UN number : 1105 Chem class : 3 Pack group : III


Quantity 1L
Packaging AcroSeal™ Glass Bottle

 View more versions of this product

Product Code. 13167714

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3-Methyl-1-butanol
98.5
99%
C5H12O
88.15
123-51-3
100.0
99%
(CH3)2CHCH2CH2OH
Isoamyl alcohol, Isopentyl alcohol

Specifications

3-Methyl-1-butanol
98.5
0.8000g/mL
AcroSeal™ Glass Bottle
1.4050 to 1.4070
99%
C5H12O
0.8
0.005% Max.
− 70.0°C
130.0°C
1L
123-51-3
100.0
Authentic
Liquid
99%
0.0005 % max.
(CH3)2CHCH2CH2OH
88.15
Isoamyl alcohol, Isopentyl alcohol
Clear colorless
45°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Methyl-1-butanol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS