missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,2-dihydropyridin-2-one, 97%, Thermo Scientific™

Catalog Number p-4391399
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
1003-56-1
C6H7NO
109.128
MVKDNXIKAWKCCS-UHFFFAOYSA-N
3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone
70481
3-methyl-1H-pyridin-2-one
CC1=CC=CNC1=O
This item is not returnable. View return policy
1003-56-1
C6H7NO
109.128
MVKDNXIKAWKCCS-UHFFFAOYSA-N
3-methylpyridin-2-ol, 2-hydroxy-3-methylpyridine, 3-methyl-2-pyridone, 3-methylpyridin-2 1h-one, 3-methyl-2-pyridinol, 2 1h-pyridinone, 3-methyl, 3-methyl-2-pyridinone, unii-vm19fn4hhv, 3-methyl-2-hydroxypyridine, 3-methyl-2 1h-pyridinone
70481
3-methyl-1H-pyridin-2-one
CC1=CC=CNC1=O
Assay Percent Range 97%
Quantity 1 g
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 3-Methyl-1, 2-dihydropyridin-2-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'