missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1,2-cyclopentanedione, 99%, Thermo Scientific Chemicals

Catalog Number p-3779075
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
765-70-8
C6H8O2
112.13
MFCD00001417
OACYKCIZDVVNJL-UHFFFAOYSA-N
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
61209
3-methylcyclopentane-1,2-dione
CC1CCC(=O)C1=O
This item is not returnable. View return policy
765-70-8
C6H8O2
112.13
MFCD00001417
OACYKCIZDVVNJL-UHFFFAOYSA-N
3-methyl-1,2-cyclopentanedione, 1,2-cyclopentanedione, 3-methyl, methylcyclopentenolone diketo form, fema no. 2700, methylcyclopentenolone natural, kentonarome, benzil-related compound, 45, 3-methyl-1,2-cyclopentanedione maple lactone, maple lactone mcp, methyl-cyclopentenolone
61209
3-methylcyclopentane-1,2-dione
CC1CCC(=O)C1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 105.0°C to 110.0°C
Color White
Density 100g/mL
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 07, I, 310
Solubility Information Solubility in water: soluble.
Formula Weight 112.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Methyl-1,2-cyclopentanedione
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'