missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxythiophenol, 97%, Thermo Scientific Chemicals
18.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-Methoxythiophenol is used in the preparation of allenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methoxythiophenol is used in the preparation of allenes.
Solubility
Not miscible or difficult to mix in water. Soluble in methanol, benzene, hexane, toluene and dichloromethane.
Notes
Store under dry inert gas. It is sensitive to air. Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
3-Methoxythiophenol is used in the preparation of allenes.
Solubility
Not miscible or difficult to mix in water. Soluble in methanol, benzene, hexane, toluene and dichloromethane.
Notes
Store under dry inert gas. It is sensitive to air. Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Density | 1.136 |
Boiling Point | 225°C to 227°C |
Flash Point | 96°C (204°F) |
Odor | Unpleasant |
Refractive Index | 1.5865 |
Quantity | 1 g |
Beilstein | 2041496 |
Sensitivity | Air Sensitive |
Solubility Information | Not miscible or difficult to mix in water. Soluble in methanol,benzene,hexane,toluene and dichloromethane. |
Formula Weight | 140.21 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'