missing translation for 'onlineSavingsMsg'
Learn More

3-methoxythiophene, ACROS Organics™

Brand:  Acros Organics 381680050

Code : 29

Additional Details : CAS Number : 17593-92-1 Weight : 0.05500kg Transport : UN number : 1993 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10548663

  • £57.00 / 5g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

3-Methoxythiophene
97.5
98%
C5H6OS
MFCD00043894
17573-92-1
100.0
98%
114.17

Specifications

3-Methoxythiophene
97.5
Glass bottle
1.5300 to 1.5320 (20°C, 589nm)
Liquid
98%
C5H6OS
MFCD00043894
Clear light brown
49°C
17573-92-1
100.0
Authentic
1.143
98%
1.1430g/mL
114.17
80.0° to 82.0°C (65.0 mmHg)
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Methoxythiophene

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS