missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals
12.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 mL
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Melting Point | -65°C |
Color | Colorless |
Density | 0.8700g/mL |
Boiling Point | 118°C |
Flash Point | 48°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- 3-Methoxypropylamine
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 A
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'