missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxypropyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 100 g |
Formula Weight | 131.2 |
Percent Purity | 97% |
Chemical Name or Material | 3-Methoxypropyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'