Learn More
3-Methoxypropyl isothiocyanate, 97%, Maybridge™
Brand: Maybridge TL04220EE
Code : 31
Additional Details : CAS Number : 17702-11-3 Weight : 0.10000kg Transport : UN number : 3265 Chem class : 8 Pack group : II
Quantity | 50g |
---|
Chemical Identifiers
17702-11-3 | |
131.193 | |
UKVFWFDXYBLOLU-UHFFFAOYSA-N | |
140274 | |
COCCCN=C=S |
C5H9NOS | |
MFCD00022063 | |
3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, # | |
1-isothiocyanato-3-methoxypropane |
Specifications
3-Methoxypropyl isothiocyanate | |
17702-11-3 | |
97% | |
MFCD00022063 | |
UKVFWFDXYBLOLU-UHFFFAOYSA-N | |
1-isothiocyanato-3-methoxypropane | |
140274 | |
97% |
50g | |
97.0 | |
C5H9NOS | |
3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, # | |
COCCCN=C=S | |
131.193 | |
131.2 |