missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetylene, 96%, Thermo Scientific Chemicals

Catalog Number p-7036831
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
768-70-7
C9H8O
132.16
MFCD00160810
ZASXCTCNZKFDTP-UHFFFAOYSA-N
3-ethynylanisole, 3-methoxyphenylacetylene, 1-ethynyl-3-methoxy-benzene, benzene, 1-ethynyl-3-methoxy, 3-ethynyl-1-methoxybenzene, 1-etynyl-3-methoxy-benzene, 3-ethynyl-anisole, m-methoxyphenylacetylene, 3-ethynylanisole 96
640753
1-ethynyl-3-methoxybenzene
COC1=CC=CC(=C1)C#C
This item is not returnable. View return policy
768-70-7
C9H8O
132.16
MFCD00160810
ZASXCTCNZKFDTP-UHFFFAOYSA-N
3-ethynylanisole, 3-methoxyphenylacetylene, 1-ethynyl-3-methoxy-benzene, benzene, 1-ethynyl-3-methoxy, 3-ethynyl-1-methoxybenzene, 1-etynyl-3-methoxy-benzene, 3-ethynyl-anisole, m-methoxyphenylacetylene, 3-ethynylanisole 96
640753
1-ethynyl-3-methoxybenzene
COC1=CC=CC(=C1)C#C

It is a useful aromatic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful aromatic compound.

Solubility
Slightly soluble in water.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.04
Boiling Point 204°C to 210°C
Flash Point 76°C (169°F)
Refractive Index 1.555
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 132.16
Percent Purity 96%
Chemical Name or Material 3-Methoxyphenylacetylene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'