missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog Number 10450111
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18908-07-1
C8H7NO2
149.15
MFCD00002019
NPOVTGVGOBJZPY-UHFFFAOYSA-N
3-methoxyphenyl isocyanate, benzene, 1-isocyanato-3-methoxy, isocyanic acid 3-methoxyphenyl ester, 3-isocyanatoanisole, 3-methoxyphenylisocyanate, m-methoxyphenyl isocyanate, 1-isocyanato-3-methoxy-benzene, 3-methoxybenzenisocyanate, m-anisyl isocyanate, m-methoxyphenylisocyanate
87843
1-isocyanato-3-methoxybenzene
COC1=CC=CC(=C1)N=C=O
This item is not returnable. View return policy
18908-07-1
C8H7NO2
149.15
MFCD00002019
NPOVTGVGOBJZPY-UHFFFAOYSA-N
3-methoxyphenyl isocyanate, benzene, 1-isocyanato-3-methoxy, isocyanic acid 3-methoxyphenyl ester, 3-isocyanatoanisole, 3-methoxyphenylisocyanate, m-methoxyphenyl isocyanate, 1-isocyanato-3-methoxy-benzene, 3-methoxybenzenisocyanate, m-anisyl isocyanate, m-methoxyphenylisocyanate
87843
1-isocyanato-3-methoxybenzene
COC1=CC=CC(=C1)N=C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Pink
Density 1.1300g/mL
Boiling Point 94°C to 95°C (10.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4NCO
Refractive Index 1.5420 to 1.544
Quantity 100 g
Beilstein 13, III, 963
Specific Gravity 1.13
Formula Weight 149.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methoxyphenyl isocyanate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methoxyphenyl isocyanate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'