missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenol, 97%, ACROS Organics™

Brand:  Acros Organics 161065000

Code : 29

Additional Details : CAS Number : 150-19-6 Weight : 0.62250kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III


Quantity 500mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10468350

  • £189.80 / 500mL


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

3-Methoxyphenol
96.0
97%
C7H8O2
124.14
m-Hydroxyanisole
150-19-6
100.0
97%
CH3OC6H4OH
MFCD00002267

Specifications

3-Methoxyphenol
>110°C
Clear pink-reddish to brown
500mL
96.0
1.5510 to 1.5530
Glass bottle
97%
C7H8O2
124.14
06, 813
m-Hydroxyanisole
1.1450g/mL
244.0°C
− 17.0°C
150-19-6
100.0
Authentic
Liquid
97%
CH3OC6H4OH
MFCD00002267
13,181
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Methoxyphenol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS