missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenethylamine, 98+%, ACROS Organics™

Brand:  Acros Organics 302610050

UNSPSC : 12350000

Code : X2

Additional Details : CAS Number : 2039-67-0 Weight : 0.05500kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10405472

  • £58.00 / 5g


Estimated Shipment
13-09-2017
Add to basket

Specifications

Chemical Identifiers

3-Methoxyphenethylamine
98.0
≥98.0%
C9H13NO
MFCD00008187
2039-67-0
100.0
98+%
151.21
2-(3-Methoxyphenyl)ethylamine

Specifications

3-Methoxyphenethylamine
Clear colorless to yellow
2039-67-0
100.0
Authentic
Liquid
98+%
151.21
13, IV, 1781
1.038g/mL
5g
98.0
1.5375 to 1.5395 (20°C, 589nm)
Glass bottle
≥98.0%
C9H13NO
MFCD00008187
2-(3-Methoxyphenyl)ethylamine
Documentation

Technical Documents

Alternative Products