missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxycarbonylphenylboronic acid, 97%, ACROS Organics™

Brand:  Acros Organics 369830250

Code : 29

Additional Details : CAS Number : 99769-19-4 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10617944

  • £449.20 / 25g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

3-Methoxycarbonylphenylboronic acid
96.0
≥96%
C8H9BO4
MFCD02093046
99769-19-4
100.0
97%
179.97

Specifications

3-Methoxycarbonylphenylboronic acid
25g
96.0
Authentic
≥96%
C8H9BO4
MFCD02093046
White to off-white
99769-19-4
100.0
Powder
97%
179.97
Glass bottle
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Methoxycarbonylphenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS