missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[Methoxy(methyl)carbamoyl]benzeneboronic acid, 97%, Alfa Aesar™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 66°C to 70°C |
Quantity | 250 mg |
Formula Weight | 209.01 |
Percent Purity | 97% |
Chemical Name or Material | 3-[Methoxy(methyl)carbamoyl]benzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'