missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™

Catalog Number p-7052506
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
447-93-8
C9H6F3NO
201.148
MFCD13185685
QASUCJMIPAXDOB-UHFFFAOYSA-N
3-methoxy-4-trifluoromethyl benzonitrile, 3-methoxy-4-trifluoromethyl-benzonitrile, benzonitrile, 3-methoxy-4-trifluoromethyl
22625191
3-methoxy-4-(trifluoromethyl)benzonitrile
COC1=C(C=CC(=C1)C#N)C(F)(F)F
This item is not returnable. View return policy
447-93-8
C9H6F3NO
201.148
MFCD13185685
QASUCJMIPAXDOB-UHFFFAOYSA-N
3-methoxy-4-trifluoromethyl benzonitrile, 3-methoxy-4-trifluoromethyl-benzonitrile, benzonitrile, 3-methoxy-4-trifluoromethyl
22625191
3-methoxy-4-(trifluoromethyl)benzonitrile
COC1=C(C=CC(=C1)C#N)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 52°C to 55°C
Color Yellow
Quantity 1 g
UN Number UN3439
Formula Weight 201.15
Percent Purity 97%
Physical Form Fused Solid
Chemical Name or Material 3-Methoxy-4-(trifluoromethyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'