missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-1,2-propanediol, 98%, Thermo Scientific Chemicals

Catalog Number p-3758336
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
623-39-2
C4H10O3
106.12
MFCD00004719
PSJBSUHYCGQTHZ-UHFFFAOYNA-N
3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
92822
3-methoxypropane-1,2-diol
COCC(O)CO
This item is not returnable. View return policy
623-39-2
C4H10O3
106.12
MFCD00004719
PSJBSUHYCGQTHZ-UHFFFAOYNA-N
3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
92822
3-methoxypropane-1,2-diol
COCC(O)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1200g/mL
Boiling Point 133°C (35.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H3COCH2CH(OH)CH2OH
Refractive Index 1.4430 to 1.445
Quantity 1 g
Specific Gravity 1.12
Solubility Information Solubility in water: soluble in water
Formula Weight 106.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1, 2-propanediol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'