missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptopropionic acid, 99+%, ACROS Organics™

Catalog Number p-155472
15.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 mL
500 mL
2.5 L
107-96-0
C3H6O2S
106.14
MFCD00004897
DKIDEFUBRARXTE-UHFFFAOYSA-N
3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid
6514
CHEBI:44111
3-sulfanylpropanoic acid
C(CS)C(=O)O
107-96-0
C3H6O2S
106.14
MFCD00004897
DKIDEFUBRARXTE-UHFFFAOYSA-N
3-mercaptopropionic acid, 3-mercaptopropanoic acid, 3-thiopropionic acid, 3-thiopropanoic acid, propanoic acid, 3-mercapto, beta-mercaptopropionic acid, 3mpa, mercaptopropionic acid, beta-thiopropionic acid, 2-mercaptoethanecarboxylic acid
6514
CHEBI:44111
3-sulfanylpropanoic acid
C(CS)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 17.0°C to 19.0°C
Color White
Density 1.2180g/mL
Boiling Point 110.0°C to 111.0°C (15.0 mmHg)
Flash Point 124°C
Infrared Spectrum Authentic
Assay Percent Range 99.0 to 101% (Iodimetry)
Packaging Glass bottle
Linear Formula HSCH2CH2CO2H
Refractive Index 1.4910 to 1.4930
Quantity 100 mL
Beilstein 03,299
Specific Gravity 1.218
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 106.14
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 3-Mercaptopropionic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Mercaptopropionic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'