missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptophenol, 96%, Thermo Scientific Chemicals

Catalog Number p-3759064
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
40248-84-8
C6H6OS
126.17
MFCD01317829
DOFIAZGYBIBEGI-UHFFFAOYSA-N
3-hydroxythiophenol, 3-mercaptophenol, 3-hydroxybenzenethiol, phenol, 3-mercapto, m-hydroxy thiophenol, 3-hydroxy thiophenol, thioresorcin, 3-hydroxythiophenol 3-mercaptophenol, 3-sulphanylphenol, 3-mercapto phenol
2736613
3-sulfanylphenol
OC1=CC(S)=CC=C1
This item is not returnable. View return policy
40248-84-8
C6H6OS
126.17
MFCD01317829
DOFIAZGYBIBEGI-UHFFFAOYSA-N
3-hydroxythiophenol, 3-mercaptophenol, 3-hydroxybenzenethiol, phenol, 3-mercapto, m-hydroxy thiophenol, 3-hydroxy thiophenol, thioresorcin, 3-hydroxythiophenol 3-mercaptophenol, 3-sulphanylphenol, 3-mercapto phenol
2736613
3-sulfanylphenol
OC1=CC(S)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6300g/mL
Boiling Point 135°C (35.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula SHC6H4OH
Quantity 1 g
Specific Gravity 1.63
Formula Weight 126.18
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Mercaptophenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Mercaptophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'