missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1-hexanol, 98%, ACROS Organics™

Catalog Number p-155464
28.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
51755-83-0
C6H14OS
134.23
MFCD00792515
TYZFMFVWHZKYSE-UHFFFAOYSA-N
3-mercapto-1-hexanol, 3-mercaptohexanol, 3-mercaptohexan-1-ol, 1-hexanol, 3-mercapto, 3-thiohexan-1-ol, 3-sulphanylhexan-1-ol, 3-sulfanyl-1-hexanol, 3-thiohexanol, acmc-209kvm, 3-mercapto-hexan-1-ol
521348
CHEBI:77690
3-sulfanylhexan-1-ol
CCCC(CCO)S
51755-83-0
C6H14OS
134.23
MFCD00792515
TYZFMFVWHZKYSE-UHFFFAOYSA-N
3-mercapto-1-hexanol, 3-mercaptohexanol, 3-mercaptohexan-1-ol, 1-hexanol, 3-mercapto, 3-thiohexan-1-ol, 3-sulphanylhexan-1-ol, 3-sulfanyl-1-hexanol, 3-thiohexanol, acmc-209kvm, 3-mercapto-hexan-1-ol
521348
CHEBI:77690
3-sulfanylhexan-1-ol
CCCC(CCO)S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 250°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H3C(CH2)2CH(SH)(CH2)2OH
Refractive Index 1.4790 to 1.481
Quantity 5 mL
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble in water.,soluble in organic solvents (most)
Formula Weight 134.23
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Mercapto-1-hexanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Mercapto-1-hexanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'