missing translation for 'onlineSavingsMsg'
Learn More

3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%, Thermo Scientific Chemicals

Catalog Number 15344597
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy
55750-62-4
C11H10N2O6
266.21
JKHVDAUOODACDU-UHFFFAOYSA-N
n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester
4620597
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate
C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O
This item is not returnable. View return policy
55750-62-4
C11H10N2O6
266.21
JKHVDAUOODACDU-UHFFFAOYSA-N
n-succinimidyl 3-maleimidopropionate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoate, 3-maleimido propionic acid n-hydroxysuccinimide ester, bmps, 3-maleimidopropionic acid n-succinimidyl ester, 3-maleimidopropionic acid nhs, succinimido 3-maleimidopropanoate, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl propanoate, 3-maleimidopropionic acid n-hydroxysuccinimide ester
4620597
(2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)propanoate
C1CC(=O)N(C1=O)OC(=O)CCN2C(=O)C=CC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 159.0°C to 160.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 100 mg
Formula Weight 266.21
Percent Purity 99%
Chemical Name or Material 3-Maleimidopropionic acid N-hydroxysuccinimide ester
compliance-icons
Product Identifier
  • 3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'