missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%, Thermo Scientific Chemicals
44.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 159.0°C to 160.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (HPLC) |
Quantity | 100 mg |
Formula Weight | 266.21 |
Percent Purity | 99% |
Chemical Name or Material | 3-Maleimidopropionic acid N-hydroxysuccinimide ester |
Safety and Handling
Product Identifier
- 3-Maleimidopropionic acid N-hydroxysuccinimide ester, 99%
- Warning
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'