missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Isopropyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63464.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15410179

  • £119.78 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Isopropyl-2-thiohydantoin
158.22
MFCD00105550
3-Isopropyl-2-thioxo-4-imidazolidinone

Specifications

3-Isopropyl-2-thiohydantoin
Solid
158.22
3-Isopropyl-2-thioxo-4-imidazolidinone
1g
95%
MFCD00105550
Videos
SDS