missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Isopropyl-1-methyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63922.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15480889

  • £109.18 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Isopropyl-1-methyl-2-thiohydantoin
172.25
MFCD27976892
3-Isopropyl-1-methyl-2-thioxo-4-imidazolidinone

Specifications

3-Isopropyl-1-methyl-2-thiohydantoin
Solid
172.25
3-Isopropyl-1-methyl-2-thioxo-4-imidazolidinone
1g
95%
MFCD27976892
Videos
SDS