missing translation for 'onlineSavingsMsg'
Learn More

3-iodotetrahydrofuran, 97%, Thermo Scientific™

Catalog Number p-3881449
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
121138-01-0
C4H7IO
198.00
MFCD09878873
BKIQORJIKOPRCG-UHFFFAOYNA-N
3-iodotetrahydrofuran, 3-iodo-tetrahydro-furan, furan, tetrahydro-3-iodo, 3-iodanyloxolane, iodotetrahydrofuran
22139264
3-iodooxolane
IC1CCOC1
This item is not returnable. View return policy
121138-01-0
C4H7IO
198.00
MFCD09878873
BKIQORJIKOPRCG-UHFFFAOYNA-N
3-iodotetrahydrofuran, 3-iodo-tetrahydro-furan, furan, tetrahydro-3-iodo, 3-iodanyloxolane, iodotetrahydrofuran
22139264
3-iodooxolane
IC1CCOC1
CAS Min % 96.5
Color Colorless
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 198
Percent Purity 97%
Boiling Point 110°C
Quantity 1 g
Packaging Amber Glass Bottle
Physical Form Liquid
Chemical Name or Material 3-Iodotetrahydrofuran
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'