missing translation for 'onlineSavingsMsg'
Learn More

3-Iodophenol, 99%, Thermo Scientific Chemicals

Catalog Number 10612135
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
626-02-8
C6H5IO
220
MFCD00002261
FXTKWBZFNQHAAO-UHFFFAOYSA-N
m-iodophenol, phenol, 3-iodo, m-hydroxyiodobenzene, 3-jodphenol, phenol, m-iodo, 3-jodphenol german, m-jodphenol, 3-iodo-phenol, 5-iodophenol, pubchem3968
12272
CHEBI:33439
3-iodophenol
C1=CC(=CC(=C1)I)O
This item is not returnable. View return policy
626-02-8
C6H5IO
220
MFCD00002261
FXTKWBZFNQHAAO-UHFFFAOYSA-N
m-iodophenol, phenol, 3-iodo, m-hydroxyiodobenzene, 3-jodphenol, phenol, m-iodo, 3-jodphenol german, m-jodphenol, 3-iodo-phenol, 5-iodophenol, pubchem3968
12272
CHEBI:33439
3-iodophenol
C1=CC(=CC(=C1)I)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 38.0°C to 43.0°C
Color Beige-Yellow to Gray
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4OH
Quantity 5 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in aqueous alkali,soluble in most common organic solvents
Formula Weight 220
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 3-Iodophenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Iodophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'