missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzotrifluoride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081709
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
401-81-0
C7H4F3I
272.009
MFCD00001049
IGISPMBUGPHLBY-UHFFFAOYSA-N
1-iodo-3-trifluoromethyl benzene, 3-iodobenzotrifluoride, m-iodobenzotrifluoride, benzene, 1-iodo-3-trifluoromethyl, 3-iodobenztrifluoride, m-trifluoromethyl iodobenzene, 3-iodo-alpha,alpha,alpha-trifluorotoluene, 3-trifluoromethyliodobenzene, 3-trifluoromethyl-iodobenzene, alpha,alpha,alpha-trifluoro-m-iodotoluene
67868
1-iodo-3-(trifluoromethyl)benzene
C1=CC(=CC(=C1)I)C(F)(F)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
401-81-0
C7H4F3I
272.009
MFCD00001049
IGISPMBUGPHLBY-UHFFFAOYSA-N
1-iodo-3-trifluoromethyl benzene, 3-iodobenzotrifluoride, m-iodobenzotrifluoride, benzene, 1-iodo-3-trifluoromethyl, 3-iodobenztrifluoride, m-trifluoromethyl iodobenzene, 3-iodo-alpha,alpha,alpha-trifluorotoluene, 3-trifluoromethyliodobenzene, 3-trifluoromethyl-iodobenzene, alpha,alpha,alpha-trifluoro-m-iodotoluene
67868
1-iodo-3-(trifluoromethyl)benzene
C1=CC(=CC(=C1)I)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -8°C
Density 1.882
Boiling Point 81°C to 82°C (24 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.5185
Quantity 10 g
Beilstein 2088596
Sensitivity Light sensitive
Formula Weight 272.01
Percent Purity ≥98%
Chemical Name or Material 3-Iodobenzotrifluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'