missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzonitrile, 97%, ACROS Organics™

Brand:  Acros Organics 368600500

Code : 29

Additional Details : CAS Number : 69113-59-3 Weight : 0.10000kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III


Quantity 50g

 View more versions of this product

Product Code. 11945621

  • £307.00 / 50g


Call for latest availability
+44 (0)1509 555 500
Add to basket


Specifications

Chemical Identifiers

3-Iodobenzonitrile
69113-59-3
100.0
C7H4IN
MFCD00079762
97%
96.0
97%
229.02

Specifications

3-Iodobenzonitrile
69113-59-3
100.0
97%
229.02
39.0° to 44.0°C
>100°C
97%
96.0
97%
C7H4IN
MFCD00079762
50g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Iodobenzonitrile

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS