missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzoic Acid 98%, ACROS Organics™

Brand:  Acros Organics 160480500

Code : 29

Additional Details : CAS Number : 618-51-9 Weight : 0.10000kg


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10541411

  • £54.60 / 50g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

3-Iodobenzoic acid
618-51-9
100.0
98%
IC6H4CO2H
MFCD00002496
98%
97.5
≥97.5%
C7H5IO2
248.01

Specifications

3-Iodobenzoic acid
618-51-9
100.0
Glass bottle
≥97.5%
C7H5IO2
248.01
09, 365
185° to 189°C
98%
97.5
Authentic
Powder
98%
IC6H4CO2H
MFCD00002496
White to beige
50g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Iodobenzoic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS