missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodobenzoic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Soluble in Chloroform and Methanol. Insoluble in water.
Notes
Light Sensitive. Store in the dark. Incompatible materials are bases and oxidizing agents.
3-Iodobenzoic acid is used in solid phase synthesis of γ-turn mimetic library. It is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Soluble in Chloroform and Methanol. Insoluble in water.
Notes
Light Sensitive. Store in the dark. Incompatible materials are bases and oxidizing agents.
Specifications
Melting Point | 186°C to 188°C |
Quantity | 50 g |
Beilstein | 971088 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in Chloroform and Methanol. Insoluble in water. |
Formula Weight | 248.02 |
Percent Purity | ≥98% |
Chemical Name or Material | 3-Iodobenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'