missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodoaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store at 4°C. Store protected from light. Incompatible with Acid chlorides, acid anhydrides, acids, chloroformates, strong oxidizing agents.
3-Iodoanilineis used as Anilines, Aromatic Amines and Nitro Compounds.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store at 4°C. Store protected from light. Incompatible with Acid chlorides, acid anhydrides, acids, chloroformates, strong oxidizing agents.
Specifications
Melting Point | 21°C to 24°C |
Density | 1.821 |
Boiling Point | 145°C to 146°C (15 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.684 |
Quantity | 10 g |
Beilstein | 636058 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 219.03 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'