missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-L-tyrosine, 98%, Thermo Scientific Chemicals
74.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
3-iodo-l-tyrosine is used in the inhibition of tyrosine hydroxylase. 3-iodo-L-tyrosine is used in the rapid induction of prolactin secretion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-iodo-l-tyrosine is used in the inhibition of tyrosine hydroxylase. 3-iodo-L-tyrosine is used in the rapid induction of prolactin secretion.
Solubility
Soluble in dilute aqeous acid and dilute aqeous base.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
3-iodo-l-tyrosine is used in the inhibition of tyrosine hydroxylase. 3-iodo-L-tyrosine is used in the rapid induction of prolactin secretion.
Solubility
Soluble in dilute aqeous acid and dilute aqeous base.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
Specifications
Melting Point | 210°C (decomposition) |
Color | White |
Beilstein | 2941266 |
Solubility Information | Soluble in dilute aqeous acid and dilute aqeous base. |
Formula Weight | 307.09 |
Percent Purity | 98% |
Quantity | 5 g |
Sensitivity | Light sensitive |
Physical Form | Powder |
Chemical Name or Material | 3-Iodo-L-tyrosine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'