missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Iodo-9-phenylcarbazole, 98%

Brand:  Alfa Aesar™ H64926.03

Code : NEW

Additional Details : CAS Number : 502161-03-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15462349

  • £11.98 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Iodo-9-phenylcarbazole
369.21
9269252
502161-03-7
MFCD09261276
3-Iodo-N-phenylcarbazole

Specifications

3-Iodo-9-phenylcarbazole
Solid
369.21
9269252
3-Iodo-N-phenylcarbazole
502161-03-7
98%
MFCD09261276
1g
Light Sensitive
Videos
SDS